5-Nitroindole ≥98.0%

Supplier: TCI America

N0400-5G N0400-25G
TCN0400-005GEA 105.86 CAD
TCN0400-005G TCN0400-025G
5-Nitroindole ≥98.0%
5-Nitroindole

CAS Number: 6146-52-7
MDL Number: MFCD00005673
Molecular Formula: C8H6N2O2
Molecular Weight: 162.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 142

Formula: C₈H₆N₂O₂
Melting Pt: 142 °C
MDL Number: MFCD00005673
CAS Number: 6146-52-7
EINECS: 228-153-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=19769ebff70689232b3aa6e79dc0b86b210f6b28&vid=19769ebff70689232b3aa6e79dc0b86b210f6b28&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR