6-Nitro-1H-indazole ≥98.0%
Supplier: TCI America
Synonyms:
6-Nitroindazole, 6-Nitro-1H-indazole, 6-NINA
N0399-250G
N0399-25G
TCN0399-250GEA
241.17
CAD
TCN0399-250G
TCN0399-025G
6-Nitro-1H-indazole ≥98.0%
6-Nitro-1H-indazole
CAS Number: 7597-18-4
MDL Number: MFCD00005695
Molecular Formula: C7H5N3O2
Molecular Weight: 163.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 182
Formula:
C₇H₅N₃O₂ MW: 163.14 g/mol Melting Pt: 181 |
MDL Number:
MFCD00005695 CAS Number: 7597-18-4 EINECS: 231-500-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...