2,6-Naphthalenedicarboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2,6-Naphthalenedicarboxylic Acid, Naphthalene-2,6-dicarboxylic acid

N0377-25G N0377-100G N0377-5G
TCN0377-025GEA 183.28 CAD
TCN0377-025G TCN0377-100G 76220-352
2,6-Naphthalenedicarboxylic acid ≥98.0% (by GC, titration analysis)
2,6-Naphthalenedicarboxylic acid

CAS Number: 1141-38-4
MDL Number: MFCD00004105
Molecular Formula: C12H8O4
Molecular Weight: 216.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Flash Point (°C): 200

Formula: C₁₂H₈O₄
MW: 216.19 g/mol
Boiling Pt: 422
Melting Pt: 366…368 °C
Flash Pt: 200 °C
MDL Number: MFCD00004105
CAS Number: 1141-38-4
EINECS: 214-527-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1be42e26333f65d92c14a2a3f0bbc938361fa440&vid=1be42e26333f65d92c14a2a3f0bbc938361fa440&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR