4-Nitrobenzophenone ≥99.0%

Supplier: TCI America

Synonyms: p-Nitrobenzophenone

N0365-10G N0365-25G
TCN0365-010GEA 64.78 CAD
TCN0365-010G TCN0365-25G
4-Nitrobenzophenone ≥99.0%
4-Nitrobenzophenone

CAS Number: 1144-74-7
MDL Number: MFCD00007354
Molecular Formula: C13H9NO3
Molecular Weight: 227.22
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 138

Formula: C₁₃H₉NO₃
MW: 227.22 g/mol
Melting Pt: 138 °C
Density: 1.406 g/cm³ (0 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007354
CAS Number: 1144-74-7
EINECS: 214-542-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2e48ed9ec52c23ac28e7fa51781bdac57f10f252&vid=2e48ed9ec52c23ac28e7fa51781bdac57f10f252&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR