3-Nitrocinnamic acid ≥98.0%

Supplier: TCI America

Synonyms: 3-(3-Nitrophenyl)acrylic acid

N0354-25G N0354-5G
TCN0354-025GEA 86.79 CAD
TCN0354-025G TCN0354-5G
3-Nitrocinnamic acid ≥98.0%
3-Nitrocinnamic acid

CAS Number: 555-68-0
MDL Number: MFCD00007277
Molecular Formula: C9H7NO4
Molecular Weight: 193.16
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 205

Formula: C₉H₇NO₄
Melting Pt: 205 °C
MDL Number: MFCD00007277
CAS Number: 555-68-0
EINECS: 209-104-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d6e6ff6eb7f78ee834ecdc03ed5e2d7c35aad92f&vid=d6e6ff6eb7f78ee834ecdc03ed5e2d7c35aad92f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR