2-Naphthylacetic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-Naphthaleneacetic acid, 2-(Naphthalen-2-yl)acetic acid

N0352-5G N0352-25G
TCN0352-005GEA 65.92 CAD
TCN0352-005G TCN0352-25G
2-Naphthylacetic acid ≥98.0% (by GC, titration analysis)
2-Naphthylacetic acid

CAS Number: 581-96-4
MDL Number: MFCD00004126
Molecular Formula: C12H10O2
Molecular Weight: 186.21
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 142


Other Product Names / Synonyms: 2-Naphthylacetic Acid

Formula: C₁₂H₁₀O₂
MW: 186.21 g/mol
Boiling Pt: 352 °C (1013 hPa)
Melting Pt: 142 °C
Density: 1.29 g/cm³ (20 °C)
MDL Number: MFCD00004126
CAS Number: 581-96-4
EINECS: 209-475-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a4a98e87bb3784f46e6ca8f12d2a497f5f29237&vid=5a4a98e87bb3784f46e6ca8f12d2a497f5f29237&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR