8-Nitroquinoline ≥99.0% (by GC, titration analysis)
Supplier: TCI America
N0345-25G
N0345-100G
N0345-5G
TCN0345-025GEA
238.58
CAD
TCN0345-025G
TCN0345-100G
TCN0345-5G
8-Nitroquinoline ≥99.0% (by GC, titration analysis)
8-Nitroquinoline
CAS Number: 607-35-2
MDL Number: MFCD00006806
Molecular Formula: C9H6N2O2
Molecular Weight: 174.16
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Melting point (°C): 89
Formula:
C₉H₆N₂O₂ Boiling Pt: 323 °C (1013 hPa) Melting Pt: 89 °C |
MDL Number:
MFCD00006806 CAS Number: 607-35-2 EINECS: 210-135-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...