(±)-2-Nonanol ≥98.0%

Supplier: TCI America

N0334-25ML N0334-500ML
TCN0334-025MLEA 72.68 CAD
TCN0334-025ML TCN0334-500ML
(±)-2-Nonanol ≥98.0%
(±)-2-Nonanol

CAS Number: 628-99-9
MDL Number: MFCD00004593
Molecular Formula: C9H20O
Molecular Weight: 144.26
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 198
Flash Point (°C): 82
Specific Gravity (20/20): 0.82


Other Product Names / Synonyms: Heptylmethylcarbinol, Methylheptylcarbinol, 2-Nonyl Alcohol

Formula: C₉H₂₀O
Boiling Pt: 198 °C
Melting Pt: –35 °C
Density: 0.82 (20 °C)
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00004593
CAS Number: 628-99-9
EINECS: 211-065-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb2a5d2bb440151136871a58f077fdb64a8a555d&vid=a8ca8471a3265d4b62330c7d85df69b2ae07d895&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR