D-(-)-Norleucine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

N0327-1G N0327-10G
TCN0327-001GEA 33.18 CAD
TCN0327-001G TCN0327-010G
D-(-)-Norleucine ≥98.0% (by HPLC, titration analysis)
D(-)-Norleucine

CAS Number: 327-56-0
MDL Number: MFCD00008099
Molecular Formula: C6H13NO2
Molecular Weight: 131.18
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Specific rotation [a]20/D: -23 deg (C=4.5, 6mol/L HCl)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: D-2-Aminohexanoic Acid

Formula: C₆H₁₃NO₂
Melting Pt: 301 °C
MDL Number: MFCD00008099
CAS Number: 327-56-0
EINECS: 206-320-9
Merck Index: 12,06800
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=80fb9a80177b2d4d0892ef498c213bfa7e891c1e&vid=80fb9a80177b2d4d0892ef498c213bfa7e891c1e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR