4-Nitrobenzamide ≥98.0%

Supplier: TCI America

N0323-500G N0323-25G
TCN0323-500GEA 1352.48 CAD
TCN0323-500G TCN0323-025G
4-Nitrobenzamide ≥98.0%
4-Nitrobenzamide

CAS Number: 619-80-7
MDL Number: MFCD00007994
Molecular Formula: C7H6N2O3
Molecular Weight: 166.14
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 203

Formula: C₇H₆N₂O₃
Melting Pt: 203 °C
Density: 1.59 g/cm³ (20 °C)
MDL Number: MFCD00007994
CAS Number: 619-80-7
EINECS: 210-613-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4855ad8d09207894c2b53a180f76c61c7d4d99ee&vid=4855ad8d09207894c2b53a180f76c61c7d4d99ee&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR