3-Nitrobenzamide ≥98.0%

Supplier: TCI America

N0321-25G N0321-500G
TCN0321-025GEA 99.54 CAD
TCN0321-025G TCN0321-500G
3-Nitrobenzamide ≥98.0%
3-Nitrobenzamide

CAS Number: 645-09-0
MDL Number: MFCD00007984
Molecular Formula: C7H6N2O3
Molecular Weight: 166.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 143

Formula: C₇H₆N₂O₃
MW: 166.14 g/mol
Boiling Pt: 310
Melting Pt: 143 °C
MDL Number: MFCD00007984
CAS Number: 645-09-0
EINECS: 211-431-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f7eb84dfb20ca210025d88a3616616963b1fd1f7&vid=f7eb84dfb20ca210025d88a3616616963b1fd1f7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR