Pelargonaldehyde ≥95.0%

Supplier: TCI America

Synonyms: 1-Nonanal, Nonylaldehyde, Nonanal, Nonaldehyde

N0296-500ML N0296-25ML N0296-100ML
TCN0296-500MLEA 268.6 CAD
TCN0296-500ML TCN0296-025ML TCN0296-100ML
Pelargonaldehyde ≥95.0%
Pelargonaldehyde

CAS Number: 124-19-6
MDL Number: MFCD00007030
Molecular Formula: C9H18O
Molecular Weight: 142.24
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 192
Flash Point (°C): 71
Specific Gravity (20/20): 0.83


Other Product Names / Synonyms: Nonanaldehyde, Nonyl Aldehyde, Pelargonaldehyde

Formula: C₉H₁₈O
MW: 142.24 g/mol
Boiling Pt: 190
Melting Pt: -40
Density: 0.83 (20 °C)
Flash Pt: 71 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007030
CAS Number: 124-19-6
EINECS: 204-688-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9054ec2591a1549225287a72f4328c0c22d81071&vid=bed93227f80405d0b83bb1b3c4c83dde1b2c62e8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR