2-Nonanone ≥98.0%

Supplier: TCI America

N0293-25ML N0293-500ML
TCN0293-025MLEA 33.73 CAD
TCN0293-025ML TCN0293-500ML
2-Nonanone ≥98.0%
2-Nonanone

CAS Number: 821-55-6
MDL Number: MFCD00009553
Molecular Formula: C9H18O
Molecular Weight: 142.24
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 195
Melting point (°C): -21
Flash Point (°C): 76
Specific Gravity (20/20): 0.82


Other Product Names / Synonyms: Heptyl Methyl Ketone

Formula: C₉H₁₈O
Boiling Pt: 195 °C
Melting Pt: –21 °C
Density: 0.82 (20 °C)
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00009553
CAS Number: 821-55-6
EINECS: 212-480-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=02ec475f5b4a1d209cd3347aa99b5b6210d6d40b&vid=83377898df430098b9380180afe44fa04d6f87b4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR