Ethyl pelargonate ≥95.0%
Supplier: TCI America
Synonyms:
Ethyl nonanoate, Pelargonic acid ethyl ester, Nonanoic acid ethyl ester
CAS Number: 123-29-5
MDL Number: MFCD00009570
Molecular Formula: C11H22O2
Molecular Weight: 186.30
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 220
Melting point (°C): -44
Flash Point (°C): 104
Specific Gravity (20/20): 0.87
Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Ethyl Pelargonate, Nonanoic Acid Ethyl Ester, Nonylic Acid Ethyl Ester, Pelargonic Acid Ethyl Ester
Formula:
C₁₁H₂₂O₂ MW: 186.29 g/mol Boiling Pt: 220 °C Melting Pt: –44 °C Density: 0.87 (20 °C) Flash Pt: 104 °C |
MDL Number:
MFCD00009570 CAS Number: 123-29-5 EINECS: 204-615-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...