1,9-Nonanediol ≥98.0%
Supplier: TCI America
N0287-25G
N0287-500G
N0287-100G
TCN0287-025GEA
75.84
CAD
TCN0287-025G
TCN0287-500G
TCN0287-100G
1,9-Nonanediol ≥98.0%
CAS Number: 3937-56-2
MDL Number: MFCD00002991
Molecular Formula: C9H20O2
Molecular Weight: 160.26
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 171
Melting point (°C): 46
Flash Point (°C): 162
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,9-Dihydroxynonane, Nonamethylene Glycol
Formula:
C₉H₂₀O₂ Boiling Pt: 171 °C Melting Pt: 46 °C Density: 0.918 g/cm³ (50 °C) Flash Pt: 162 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002991 CAS Number: 3937-56-2 EINECS: 223-517-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...