n-Nonadecane ≥99.0%

Supplier: TCI America

N0282-25G N0282-400G
TCN0282-025GEA 80.23 CAD
TCN0282-025G TCN0282-400G
n-Nonadecane ≥99.0%
n-Nonadecane

CAS Number: 629-92-5
MDL Number: MFCD00009012
Molecular Formula: C19H40
Molecular Weight: 268.53
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 330
Flash Point (°C): 100
Freezing point (°C): 32

Formula: H₃C(CH₂)₁₇CH₃
Boiling Pt: 330 °C
Melting Pt: 32.5 °C
Density: 0.7845 g/cm³ (20 °C)
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00009012
CAS Number: 629-92-5
EINECS: 211-116-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=beb2db320bb2a4d902c68309f42979e341972b27&vid=9857190c55df15efb7830be7421bed9f8956fc2e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR