2-Nitroso-1-naphthol ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

N0267-25G N0267-5G
TCN0267-025GEA 328.64 CAD
TCN0267-025G TCN0267-5G
2-Nitroso-1-naphthol ≥98.0% (by GC, titration analysis)
2-Nitroso-1-naphthol

CAS Number: 132-53-6
MDL Number: MFCD00003932
Molecular Formula: C10H7NO2
Molecular Weight: 173.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 148

Formula: C₁₀H₇NO₂
Boiling Pt: 304 °C (1013 hPa)
Melting Pt: 148 °C
Storage Temperature: Ambient
MDL Number: MFCD00003932
CAS Number: 132-53-6
EINECS: 205-064-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c6c69796fe09ae09d1bc34d56c2d61055da2e12a&vid=c6c69796fe09ae09d1bc34d56c2d61055da2e12a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR