5-Nitrosalicylaldehyde ≥97.0%

Supplier: TCI America
Warning

Synonyms: 2-Hydroxy-5-nitrobenzaldehyde

N0255-5G N0255-25G
TCN0255-5GEA 59.57 CAD
TCN0255-5G TCN0255-025G
5-Nitrosalicylaldehyde ≥97.0%
5-Nitrosalicylaldehyde

CAS Number: 97-51-8
MDL Number: MFCD00007337
Molecular Formula: C7H5NO4
Molecular Weight: 167.12
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 125

Formula: C₇H₅NO₄
MW: 167.12 g/mol
Melting Pt: 125 °C
Storage Temperature: Ambient
MDL Number: MFCD00007337
CAS Number: 97-51-8
EINECS: 202-587-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d3c3885f57cd66608865195ba6ab5a0bc6bf6b29&vid=01aff072f43bdf88513552ef153b69134f512e0d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR