2-Nitroresorcinol ≥99.0% (by titrimetric analysis)
Supplier: TCI America
N0254-25G
N0254-5G
TCN0254-025GEA
124.82
CAD
TCN0254-025G
TCN0254-005G
2-Nitroresorcinol ≥99.0% (by titrimetric analysis)
2-Nitroresorcinol
CAS Number: 601-89-8
MDL Number: MFCD00007124
Molecular Formula: C6H5NO4
Molecular Weight: 155.11
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Boiling point (°C): 234
Melting point (°C): 83
Formula:
C₆H₅NO₄ Boiling Pt: 234 °C Melting Pt: 83 °C Density: 1.3526 g/cm³ (96.7 °C) |
MDL Number:
MFCD00007124 CAS Number: 601-89-8 EINECS: 210-010-9 UN: 2811 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...