6-Nitroquinoline ≥98.0% (by GC, titration analysis)
Supplier: TCI America
N0252-25G
N0252-5G
TCN0252-025GEA
233.84
CAD
TCN0252-025G
TCN0252-5G
6-Nitroquinoline ≥98.0% (by GC, titration analysis)
6-Nitroquinoline
CAS Number: 613-50-3
MDL Number: MFCD00006799
Molecular Formula: C9H6N2O2
Molecular Weight: 174.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 151
Formula:
C₉H₆N₂O₂ Boiling Pt: 335 °C (1013 hPa) Melting Pt: 151 °C |
MDL Number:
MFCD00006799 CAS Number: 613-50-3 EINECS: 210-346-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...