4-Nitrophthalimide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

N0247-25G N0247-500G
TCN0247-025GEA 50.56 CAD
TCN0247-025G TCN0247-500G
4-Nitrophthalimide ≥98.0% (by HPLC, titration analysis)
4-Nitrophthalimide

CAS Number: 89-40-7
MDL Number: MFCD00005884
Molecular Formula: C8H4N2O4
Molecular Weight: 192.13
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 202

Formula: C₈H₄N₂O₄
Melting Pt: 202 °C
MDL Number: MFCD00005884
CAS Number: 89-40-7
EINECS: 201-905-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=49622268bc21b10fb7ab9f28816d3790914488ad&vid=49622268bc21b10fb7ab9f28816d3790914488ad&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR