4-Nitrophthalic anhydride ≥97.0%
Supplier: TCI America
Synonyms:
5-Nitroisobenzofuran-1,3-dione
N0246-100G
N0246-25G
TCN0246-100GEA
718.9
CAD
TCN0246-100G
TCN0246-025G
4-Nitrophthalic anhydride ≥97.0%
4-Nitrophthalic anhydride
CAS Number: 5466-84-2
MDL Number: MFCD00005922
Molecular Formula: C8H3NO5
Molecular Weight: 193.11
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Boiling point (°C): 197
Melting point (°C): 120
Formula:
C₈H₃NO₅ MW: 193.12 g/mol Boiling Pt: 197 °C Melting Pt: 120 °C |
MDL Number:
MFCD00005922 CAS Number: 5466-84-2 EINECS: 226-776-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...