3-Nitrophthalic anhydride ≥97.0%

Supplier: TCI America
Warning

N0245-25G N0245-250G
TCN0245-025GEA 51.19 CAD
TCN0245-025G TCN0245-250G
3-Nitrophthalic anhydride ≥97.0%
3-Nitrophthalic anhydride

CAS Number: 641-70-3
MDL Number: MFCD00005921
Molecular Formula: C8H3NO5
Molecular Weight: 193.11
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 162

Formula: C₈H₃NO₅
MW: 193.12 g/mol
Melting Pt: 162 °C
MDL Number: MFCD00005921
CAS Number: 641-70-3
EINECS: 211-373-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=43bdc4d6ee6548460bf6b9a617df8bfb2b2d51e0&vid=43bdc4d6ee6548460bf6b9a617df8bfb2b2d51e0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR