4-Nitrophthalic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
N0244-25G
N0244-500G
TCN0244-025GEA
85.32
CAD
TCN0244-025G
TCN0244-500G
4-Nitrophthalic acid ≥98.0% (by HPLC, titration analysis)
4-Nitrophthalic acid
CAS Number: 610-27-5
MDL Number: MFCD00007252
Molecular Formula: C8H5NO6
Molecular Weight: 211.13
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 170
Formula:
C₈H₅NO₆ Melting Pt: 170 °C |
MDL Number:
MFCD00007252 CAS Number: 610-27-5 EINECS: 210-215-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...