3-Nitrophthalic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

N0243-25G N0243-500G
TCN0243-025GEA 50.56 CAD
TCN0243-025G TCN0243-500G
3-Nitrophthalic acid ≥98.0% (by HPLC, titration analysis)
3-Nitrophthalic acid

CAS Number: 603-11-2
MDL Number: MFCD00007138
Molecular Formula: C8H5NO6
Molecular Weight: 211.13
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal

Formula: C₈H₅NO₆
Boiling Pt: 441 °C (1013 hPa)
Melting Pt: 210…215 °C
MDL Number: MFCD00007138
CAS Number: 603-11-2
EINECS: 210-030-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63795eeac3eb35983af4556c998ce7ed2b32a767&vid=63795eeac3eb35983af4556c998ce7ed2b32a767&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR