(4-Nitrophenyl)-β-D-glucopyranoside ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: p-Nitrophenyl-β-D-Glucopyranoside, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(4-nitrophenoxy)tetrahydro-2H-pyran-3,4,5-triol

N0235-5G N0235-1G
TCN0235-005GEA 396.58 CAD
TCN0235-005G TCN0235-001G
(4-Nitrophenyl)-β-D-glucopyranoside ≥98.0% (by HPLC)
(4-Nitrophenyl) β-D-glucopyranoside

[Substrate for beta-D-Glucosidase]
CAS Number: 2492-87-7
MDL Number: MFCD00006593
Molecular Formula: C12H15NO8
Molecular Weight: 301.25
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 167
Specific rotation [a]20/D: -102.5 deg (C=1, H2O)
Storage Temperature: 0-10°C

Formula: C₁₂H₁₅NO₈
MW: 301.25 g/mol
Melting Pt: 167 °C
MDL Number: MFCD00006593
CAS Number: 2492-87-7
EINECS: 219-661-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c0ff20001657ac014b0441ae6c9d28244a44f3c3&vid=680d7b0f3289bb395afa679b77efa625eda55ad5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR