3-Nitrobiphenyl ≥99.0%

Supplier: TCI America

N0193-1G N0193-25G
TCN0193-001GEA 53.72 CAD
TCN0193-001G TCN0193-025G
3-Nitrobiphenyl ≥99.0%
3-Nitrobiphenyl

CAS Number: 2113-58-8
MDL Number: MFCD00007245
Molecular Formula: C12H9NO2
Molecular Weight: 199.21
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 204
Melting point (°C): 61


Other Product Names / Synonyms: 3-Nitrobiphenyl

Formula: C₁₂H₉NO₂
Boiling Pt: 204 °C
Melting Pt: 61 °C
Density: 1.305 g/cm³ (20 °C)
MDL Number: MFCD00007245
CAS Number: 2113-58-8
EINECS: 218-305-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dcc771532b8975dc66c7b68386b44c9d5d7017dd&vid=dcc771532b8975dc66c7b68386b44c9d5d7017dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR