4-(4-Nitrobenzyl)pyridine ≥98.0%

Supplier: TCI America

Synonyms: 4-(p-Nitrobenzyl)Pyridine

N0183-25G N0183-1G
TCN0183-025GEA 158.63 CAD
TCN0183-025G TCN0183-001G
4-(4-Nitrobenzyl)pyridine ≥98.0%
4-(4-Nitrobenzyl)pyridine

CAS Number: 1083-48-3
MDL Number: MFCD00006445
Molecular Formula: C12H10N2O2
Molecular Weight: 214.22
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 72

Formula: C₁₂H₁₀N₂O₂
MW: 214.22 g/mol
Melting Pt: 72 °C
Storage Temperature: Ambient
MDL Number: MFCD00006445
CAS Number: 1083-48-3
EINECS: 214-108-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=15479a3b42d363670d300ad8c9c6b922b4391ad3&vid=15479a3b42d363670d300ad8c9c6b922b4391ad3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR