4-Nitrobenzyl alcohol ≥98.0%
Supplier: TCI America
Synonyms:
(4-Nitrophenyl)methanol
N0180-25G
N0180-100G
TCN0180-500GEA
256.24
CAD
TCN0180-500G
TCN0180-025G
TCN0180-100G
4-Nitrobenzyl alcohol ≥98.0%
4-Nitrobenzyl alcohol
CAS Number: 619-73-8
MDL Number: MFCD00007376
Molecular Formula: C7H7NO3
Molecular Weight: 153.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 185
Melting point (°C): 95
Formula:
C₇H₇NO₃ Boiling Pt: 185 °C Melting Pt: 95 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007376 CAS Number: 619-73-8 EINECS: 210-611-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...