2-Nitrobenzyl alcohol ≥98.0%

Supplier: TCI America

N0179-25G N0179-250G
TCN0179-025GEA 88.48 CAD
TCN0179-025G TCN0179-250G
2-Nitrobenzyl alcohol ≥98.0%
2-Nitrobenzyl alcohol

CAS Number: 612-25-9
MDL Number: MFCD00007186
Molecular Formula: C7H7NO3
Molecular Weight: 153.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 168
Melting point (°C): 74
Flash Point (°C): 110

Formula: C₇H₇NO₃
Boiling Pt: 168 °C
Melting Pt: 74 °C
Flash Pt: 110 °C
MDL Number: MFCD00007186
CAS Number: 612-25-9
EINECS: 210-302-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9e0f8ce89c8e8a062dcd33cfd5b0d7d42a083cc5&vid=9e0f8ce89c8e8a062dcd33cfd5b0d7d42a083cc5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR