3-Nitrobenzyl alcohol ≥97.0%
Supplier: TCI America
N0178-25G
N0178-100G
TCN0178-025GEA
60.04
CAD
TCN0178-025G
TCN0178-100G
3-Nitrobenzyl alcohol ≥97.0%
3-Nitrobenzyl alcohol
CAS Number: 619-25-0
MDL Number: MFCD00007273
Molecular Formula: C7H7NO3
Molecular Weight: 153.14
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Flash Point (°C): 110
Freezing point (°C): 29
Formula:
C₇H₇NO₃ Boiling Pt: 317 °C (1013 hPa) Melting Pt: 26…27 °C Density: 1.3 g/cm³ (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00007273 CAS Number: 619-25-0 EINECS: 210-588-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...