Methyl-3-nitrobenzoate ≥98.0%

Supplier: TCI America

Synonyms: Methyl 3-Nitrobenzoate, 3-Nitrobenzoic acid methyl ester

N0157-500G N0157-25G
TCN0157-500GEA 949.58 CAD
TCN0157-500G TCN0157-025G
Methyl-3-nitrobenzoate ≥98.0%
Methyl 3-nitrobenzoate

CAS Number: 618-95-1
MDL Number: MFCD00007250
Molecular Formula: C8H7NO4
Molecular Weight: 181.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 279
Melting point (°C): 78


Other Product Names / Synonyms: 3-Nitrobenzoic Acid Methyl Ester

Formula: C₈H₇NO₄
MW: 181.15 g/mol
Boiling Pt: 279 °C
Melting Pt: 78 °C
MDL Number: MFCD00007250
CAS Number: 618-95-1
EINECS: 210-573-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e104c157e62945b50689cc9da35f6245a19a3a3f&vid=e104c157e62945b50689cc9da35f6245a19a3a3f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR