4-Nitrobenzoic acid ≥99.0% (by GC, titration analysis)

Supplier: TCI America
Warning

N0156-25G N0156-500G
TCN0156-025GEA 47.4 CAD
TCN0156-025G TCN0156-500G
4-Nitrobenzoic acid ≥99.0% (by GC, titration analysis)
4-Nitrobenzoic acid

CAS Number: 62-23-7
MDL Number: MFCD00007352
Molecular Formula: C7H5NO4
Molecular Weight: 167.12
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Melting point (°C): 240
Flash Point (°C): 201

Formula: C₇H₅NO₄
Boiling Pt: 359 °C (1013 hPa)
Melting Pt: 240 °C
Density: 1.65 g/cm³ (20 °C)
Flash Pt: 201 °C
Storage Temperature: Ambient
MDL Number: MFCD00007352
CAS Number: 62-23-7
EINECS: 200-526-2
Merck Index: 12,06686
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6fa8fd6c4079b496184f972ed862a277dce5b17b&vid=419189e954767b7de7321110955ece51d44c0d3b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR