3-Nitrobenzoic acid ≥99.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: m-Nitrobenzoic acid

N0154-25G N0154-500G
TCN0154-025GEA 30.02 CAD
TCN0154-025G TCN0154-500G
3-Nitrobenzoic acid ≥99.0% (by GC, titration analysis)
3-Nitrobenzoic acid

CAS Number: 121-92-6
MDL Number: MFCD00007251
Molecular Formula: C7H5NO4
Molecular Weight: 167.12
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Melting point (°C): 142

Formula: C₇H₅NO₄
MW: 167.12 g/mol
Boiling Pt: 341
Melting Pt: 142 °C
Density: 1.494 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007251
CAS Number: 121-92-6
EINECS: 204-508-5
Merck Index: 12,06686
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16b2385e0e0f63e9decae00a6651bef7fbf3992a&vid=bc31ba775780116c63371a2d7b0b17995745abcf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR