Sodium 3-nitrobenzenesulfonate ≥95.0% (by titrimetric analysis)

Supplier: TCI America

N0141-25G N0141-500G
TCN0141-025GEA 31.6 CAD
TCN0141-025G TCN0141-500G
Sodium 3-nitrobenzenesulfonate ≥95.0% (by titrimetric analysis)
Sodium 3-nitrobenzenesulphonate

CAS Number: 127-68-4
MDL Number: MFCD00007490
Molecular Formula: C6H5NO5S
Molecular Weight: 225.15
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Flash Point (°C): 100


Other Product Names / Synonyms: 3-Nitrobenzenesulfonic Acid Sodium Salt

Formula: C₆H₄NNaO₅S
Flash Pt: 100 °C
MDL Number: MFCD00007490
CAS Number: 127-68-4
EINECS: 204-857-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=257c741bfef1aeba5527ba95d0f23ce2e772bbaf&vid=257c741bfef1aeba5527ba95d0f23ce2e772bbaf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR