2-Nitrobenzaldehyde ≥99.0%

Supplier: TCI America

N0130-25G N0130-500G N0130-100G
TCN0130-025GEA 100.54 CAD
TCN0130-025G TCN0130-500G TCN0130-100G
2-Nitrobenzaldehyde ≥99.0%
2-Nitrobenzaldehyde

CAS Number: 552-89-6
MDL Number: MFCD00007132
Molecular Formula: C7H5NO3
Molecular Weight: 151.12
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 236
Melting point (°C): 44

Formula: C₇H₅NO₃
Boiling Pt: 236 °C
Melting Pt: 44 °C
Density: 1.35 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007132
CAS Number: 552-89-6
EINECS: 209-025-3
Merck Index: 12,06684
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a49d25fec34193c46716fd8eef9f9f7934819c56&vid=706622d1034c6cf2ec5fb1bb3b5f4e54367172b1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR