3-Nitrobenzaldehyde ≥98.0%

Supplier: TCI America

N0129-25G N0129-500G
TCN0129-025GEA 89.59 CAD
TCN0129-025G TCN0129-500G
3-Nitrobenzaldehyde ≥98.0%
3-Nitrobenzaldehyde

CAS Number: 99-61-6
MDL Number: MFCD00007249
Molecular Formula: C7H5NO3
Molecular Weight: 151.12
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 114
Melting point (°C): 57

Formula: C₇H₅NO₃
Boiling Pt: 114 °C
Melting Pt: 57 °C
Density: 1.481 g/cm³ (0 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007249
CAS Number: 99-61-6
EINECS: 202-772-6
Merck Index: 12,06684
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e640551dfa70207e0d17b6246d1ce5215b6d1477&vid=e5dff4df3ef7a5e62367519f7ab53ec402a9a829&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR