2-Amino-5-nitrophenol ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

N0113-500G N0113-25G
TCN0113-500GEA 336.54 CAD
TCN0113-500G TCN0113-025G
2-Amino-5-nitrophenol ≥98.0% (by GC, titration analysis)
2-Amino-5-nitrophenol

CAS Number: 121-88-0
MDL Number: MFCD00007692
Molecular Formula: C6H6N2O3
Molecular Weight: 154.13
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 204


Other Product Names / Synonyms: 2-Hydroxy-4-nitroaniline

Formula: C₆H₆N₂O₃
Melting Pt: 204 °C
Density: 1.54 g/cm³ (20 °C)
MDL Number: MFCD00007692
CAS Number: 121-88-0
EINECS: 204-503-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d95609634f08e1e161cca8031ee3974ab2e709b&vid=1d95609634f08e1e161cca8031ee3974ab2e709b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR