4'-Nitroacetophenone ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
N0111-25G
N0111-500G
N0111-100G
TCN0111-025GEA
51.43
CAD
TCN0111-025G
TCN0111-500G
TCN0111-100G
4'-Nitroacetophenone ≥98.0% (by GC)
4'-Nitroacetophenone
CAS Number: 100-19-6
MDL Number: MFCD00007355
Molecular Formula: C8H7NO3
Molecular Weight: 165.15
Purity/Analysis Method:
Formula:
C₈H₇NO₃ Boiling Pt: 167 °C Melting Pt: 79 °C Flash Pt: 149 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007355 CAS Number: 100-19-6 EINECS: 202-827-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...