2'-Nitroacetophenone ≥96.0%

Supplier: TCI America
Warning

Synonyms: o-Nitroacetophenone, 1-(2-Nitrophenyl)ethanone

N0110-5G N0110-25G
TCN0110-005GEA 33.18 CAD
TCN0110-005G TCN0110-025G
2'-Nitroacetophenone ≥96.0%
2'-Nitroacetophenone

CAS Number: 577-59-3
MDL Number: MFCD00007144
Molecular Formula: C8H7NO3
Molecular Weight: 165.15
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Boiling point (°C): 179
Flash Point (°C): 110
Freezing point (°C): 25

Formula: C₈H₇NO₃
MW: 165.15 g/mol
Boiling Pt: 179 °C
Melting Pt: 23
Density: 1.24 g/cm³ (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007144
CAS Number: 577-59-3
EINECS: 209-414-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e05c451c8857def91f0ae9eda72ecc2294ac9eb6&vid=e05c451c8857def91f0ae9eda72ecc2294ac9eb6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR