3'-Nitroacetophenone ≥98.0%
Supplier: TCI America
Synonyms:
m-Nitroacetophenone
N0109-25G
N0109-100G
TCN0109-500GEA
257.86
CAD
TCN0109-500G
TCN0109-025G
TCN0109-100G
3'-Nitroacetophenone ≥98.0%
3'-Nitroacetophenone
CAS Number: 121-89-1
MDL Number: MFCD00007259
Molecular Formula: C8H7NO3
Molecular Weight: 165.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 202
Melting point (°C): 78
Formula:
C₈H₇NO₃ MW: 165.15 g/mol Boiling Pt: 202 °C Melting Pt: 78 °C Density: 1.433 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007259 CAS Number: 121-89-1 EINECS: 204-504-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...