4'-Nitroacetanilide ≥99.0%

Supplier: TCI America
Warning

N0108-500G N0108-25G
TCN0108-500GEA 1058.6 CAD
TCN0108-500G TCN0108-025G
4'-Nitroacetanilide ≥99.0%
4'-Nitroacetanilide

CAS Number: 104-04-1
MDL Number: MFCD00007303
Molecular Formula: C8H8N2O3
Molecular Weight: 180.16
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 215

Formula: C₈H₈N₂O₃
Boiling Pt: 409 °C (1013 hPa)
Melting Pt: 215 °C
Storage Temperature: Ambient
MDL Number: MFCD00007303
CAS Number: 104-04-1
EINECS: 203-169-0
Merck Index: 12,06677
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=287d258e39da1ad930f05000cb87be3c442b8f6f&vid=287d258e39da1ad930f05000cb87be3c442b8f6f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR