Triisopropanolamine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 1,1',1''-Nitrilotripropan-2-ol, 1,1',1"-Nitrilotri-2-propanol, Tris(2-hydroxypropyl)amine

N0104-25G N0104-500G
TCN0104-25GEA 74.26 CAD
TCN0104-25G TCN0104-500G
Triisopropanolamine ≥98.0% (by titrimetric analysis)
Triisopropanolamine

CAS Number: 122-20-3
MDL Number: MFCD00004533
Molecular Formula: C9H21NO3
Molecular Weight: 191.27
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Boiling point (°C): 305
Flash Point (°C): 160


Other Product Names / Synonyms: Nitrilotripropanol

Formula: C₉H₂₁NO₃
MW: 191.27 g/mol
Boiling Pt: 305 °C
Flash Pt: 160 °C
MDL Number: MFCD00004533
CAS Number: 122-20-3
EINECS: 204-528-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=959c9e2bfcf47c52d06220814f9f28b80b96f606&vid=959c9e2bfcf47c52d06220814f9f28b80b96f606&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR