Nickel(II) acetylacetonate hydrate ≥98.0%

Supplier: TCI America

Synonyms: Acetylacetone nickel(II) salt hydrate, Nickel (II) bis(2,4-pentanedionato-O,O') hydrate, Bis(2,4-pentanedionato-O,O')nickel (II) hydrate, Nickel (II) 2,4-pentanedionate hydrate, Bis(2,4-pentanedionato)nickel(II) hydrate

N0096-500G N0096-25G N0096-100G
TCN0096-500GEA 1254.52 CAD
TCN0096-500G TCN0096-25G TCN0096-100G
Nickel(II) acetylacetonate hydrate ≥98.0%
Nickel (II) acetylacetonate hydrate

CAS Number: 3264-82-2
MDL Number: MFCD00000024
Molecular Formula: C10H14NiO4
Molecular Weight: 256.91
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Activation Reaction of Unactivated Bonds, Cross-coupling Reaction using Transition Metal Catalysts
Other Product Names / Synonyms: Acetylacetone Nickel(II) Salt, Nickel(II) Acetylacetonate

Formula: C₁₀H₁₄NiO₄·xH₂O
MDL Number: MFCD00149058
CAS Number: 120156-44-7
EINECS: 221-875-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9894e7136d730b601f06248e73cbc3faa619fc7e&vid=9894e7136d730b601f06248e73cbc3faa619fc7e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR