Methyl nicotinate ≥98.0%
Supplier: TCI America
N0086-25G
N0086-500G
TCN0086-025GEA
60.12
CAD
TCN0086-025G
TCN0086-500G
Methyl nicotinate ≥98.0%
Methyl nicotinate
CAS Number: 93-60-7
MDL Number: MFCD00006388
Molecular Formula: C7H7NO2
Molecular Weight: 137.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 41
Flash Point (°C): 96
Storage Temperature: 0-10°C
Other Product Names / Synonyms: Nicotinic Acid Methyl Ester
Formula:
C₇H₇NO₂ Boiling Pt: 209 °C (1013 hPa) Melting Pt: 41 °C Density: 1.0048 g/cm³ (20 °C) Flash Pt: 96 °C |
MDL Number:
MFCD00006388 CAS Number: 93-60-7 EINECS: 202-261-8 Merck Index: 12,06174 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...