Ethyl nicotinate ≥99.0%
Supplier: TCI America
N0085-25ML
N0085-500ML
N0085-100ML
TCN0085-025MLEA
58.46
CAD
TCN0085-025ML
TCN0085-500ML
TCN0085-100ML
Ethyl nicotinate ≥99.0%
Ethyl nicotinate
CAS Number: 614-18-6
MDL Number: MFCD00006389
Molecular Formula: C8H9NO2
Molecular Weight: 151.17
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 224
Flash Point (°C): 93
Freezing point (°C): 9
Specific Gravity (20/20): 1.11
Other Product Names / Synonyms: Nicotinic Acid Ethyl Ester, Ethyl 3-Pyridinecarboxylate, 3-Pyridinecarboxylic Acid Ethyl Ester
Formula:
C₈H₉NO₂ Boiling Pt: 224 °C Density: 1.11 (20 °C) Flash Pt: 93 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006389 CAS Number: 614-18-6 EINECS: 210-370-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...