Nerol ≥98.0%

Supplier: TCI America

N0077-500ML N0077-25ML
TCN0077-500MLEA 181.7 CAD
TCN0077-500ML TCN0077-025ML
Nerol ≥98.0%
Nerol

CAS Number: 106-25-2
MDL Number: MFCD00063204
Molecular Formula: C10H18O
Molecular Weight: 154.25
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 226
Flash Point (°C): 93
Specific Gravity (20/20): 0.88

Formula: C₁₀H₁₈O
Boiling Pt: 226 °C
Density: 0.88 (20 °C)
Flash Pt: 93 °C
MDL Number: MFCD00063204
CAS Number: 106-25-2
EINECS: 203-378-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=98bca1b24d1fb185d506535701061f5b1b5b308e&vid=98bca1b24d1fb185d506535701061f5b1b5b308e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR