N-(1-Naphthyl)ethylenediamine dihydrochloride ≥98.0%

Supplier: TCI America
Warning

Synonyms: Marshall's Reagent, N-2-Aminoethyl-1-naphthylamine dihydrochloride, N-1-Naphthylethylenediamine

N0063-25G N0063-5G
TCN0063-025GEA 258.47 CAD
TCN0063-025G TCN0063-5G
N-(1-Naphthyl)ethylenediamine dihydrochloride ≥98.0%
N-2-Aminoethyl-1-naphthylamine dihydrochloride

CAS Number: 1465-25-4
MDL Number: MFCD00012556
Molecular Formula: C12H14N2
Molecular Weight: 259.17
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 190


Product Categories: Bioscience, Reagents for Nitric Oxide Research
Other Product Names / Synonyms: Bratton-Marshall Reagent, Marshall's Reagent

Formula: C₁₂H₁₆Cl₂N₂
MW: 259.18 g/mol
Melting Pt: 190 °C
Storage Temperature: Ambient
MDL Number: MFCD00012556
CAS Number: 1465-25-4
EINECS: 215-981-2
Merck Index: 12,06496
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=171b1dc26629acda30c96622c7fb409f7d18055f&vid=171b1dc26629acda30c96622c7fb409f7d18055f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR