(2-Naphthoxy)acetic acid ≥98.0%
Supplier: TCI America
Synonyms:
2-Naphthoxyacetic acid
N0045-500G
N0045-25G
TCN0045-500GEA
962.06
CAD
TCN0045-500G
TCN0045-025G
(2-Naphthoxy)acetic acid ≥98.0%
(2-Naphthoxy)acetic acid
CAS Number: 120-23-0
MDL Number: MFCD00004066
Molecular Formula: C12H10O3
Molecular Weight: 202.21
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 155
Product Categories: Bioscience, Auxins
Other Product Names / Synonyms: NOA
Formula:
C₁₂H₁₀O₃ Melting Pt: 155 °C |
MDL Number:
MFCD00004066 CAS Number: 120-23-0 EINECS: 204-380-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...