2-Naphthol ≥99.0%

Supplier: TCI America
Warning

Synonyms: 2-Hydroxynaphthalene, β-Naphthol, β-Hydroxynaphthalene

N0027-250G N0027-25G
TCN0027-250GEA 97.85 CAD
TCN0027-250G TCN0027-025G
2-Naphthol ≥99.0%
2-Naphthol

CAS Number: 135-19-3
MDL Number: MFCD00004067
Molecular Formula: C10H8O
Molecular Weight: 144.17
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 286
Melting point (°C): 121
Flash Point (°C): 153

Formula: C₁₀H₈O
MW: 144.17 g/mol
Boiling Pt: 286 °C
Melting Pt: 121 °C
Density: 1.217 g/cm³ (20 °C)
Flash Pt: 153 °C
Storage Temperature: Ambient
MDL Number: MFCD00004067
CAS Number: 135-19-3
EINECS: 205-182-7
UN: 3077
ADR: 9,III
Merck Index: 12,06471
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39fd47b639448a316bca5603e72136b283ea910c&vid=39fd47b639448a316bca5603e72136b283ea910c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR